missing translation for 'onlineSavingsMsg'
Learn More

Carbazole-2-boronic acid pinacol ester, tech. 90%, Thermo Scientific™

Catalog Number 15411758
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
871125-67-6
C18H20BNO2
293.173
MFCD07784368
RLSJGSFDSSYNPL-UHFFFAOYSA-N
9h-carbazole-2-boronic acid pinacol ester, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-9h-carbazole, 9h-carbazole,4a,9a-dihydro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, carbazole-2-boronic acid pinacol ester, 9h-carbazol-2-ylboronic acid pinacol ester, 9h-carbazol-2-yl boronic acid pinacol ester, 2-tetramethyl-1,3,2-dioxaborolan-2-yl-9h-carbazole, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-9h-carbazole, 2-9h-carbazol-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
46738013
2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole
B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=CC=CC=C4N3
This item is not returnable. View return policy
871125-67-6
C18H20BNO2
293.173
MFCD07784368
RLSJGSFDSSYNPL-UHFFFAOYSA-N
9h-carbazole-2-boronic acid pinacol ester, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-9h-carbazole, 9h-carbazole,4a,9a-dihydro-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, carbazole-2-boronic acid pinacol ester, 9h-carbazol-2-ylboronic acid pinacol ester, 9h-carbazol-2-yl boronic acid pinacol ester, 2-tetramethyl-1,3,2-dioxaborolan-2-yl-9h-carbazole, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-9h-carbazole, 2-9h-carbazol-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
46738013
2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole
B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=CC=CC=C4N3

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 293.17
Percent Purity 90%
Grade Technical
Physical Form Solid
Chemical Name or Material Carbazole-2-boronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'