Learn More
Erlab™ Captair by Erlab™ Stainless Steel Work Surfaces
Designed for laboratories specializing in fields such as biology, botany, aerospace, electronics, pharmaceutical and cosmetics. Erlab™ Captair™ Stainless Steel Work Surfaces are for use with Captair PCR work stations.
Brand:  Erlab™ I0120050001
Code : NEW
Additional Details : Weight : 11.00000kg
 View more versions of this product
Product Code. 13426583
  • £2464.50 / Each
Estimated Shipment: 07-08-2024
to see stock.
Description
Description
  • High chemical and mechanical resistance
  • Rounded corners to facilitate cleaning operations
  • Built in spill tray
Specifications
Specifications
Stainless Steel
CaptairFlow 714 ductless fume hoods