missing translation for 'onlineSavingsMsg'
Learn More

Erlab™ Captair 832 Smart Filtering Storage Cabinet

Erlab™ Captair 832 Smart Filtering Storage Cabinets provide a steady flow of air through the cabinet, capturing hazardous vapors in the filters, and returns clean air back to the room.

Brand:  Erlab™ B03TA/11/AS

Additional Details : Weight : 70.00000kg

Product Code. 15555124

  • £2495.00 / Each
Estimated Shipment: 01-08-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Air Flow: 70 m3 /h
  • Electrical Requirements: 80 to 220 V, 50/60 Hz
  • Corrosion resistant electro-galvanized steel coated with anti-acid polymer
  • Doors: Clear, chemical resistant acrylic for easy viewing
  • eGuard app (Android or iOS): For remote control to monitor the status of the storage cabinet, change the settings, and deliver safety alerts immediately to your devices (mobile, tablet and PC)
  • Alarm if doors are left open
  • Eliminate inhalation of noxious or hazardous vapors when opening the cabinet
  • Continuously purify the laboratory air
  • Requires no connection to HVAC neede, so your chemicals can be placed where you need them
  • Desgined with separate compartments to accommodate multiple types of chemicals
  • Spill-proof trays
Specifications

Specifications

Ductless Filtering Chemical Storage Cabinets
500 x 650 x 2050 mm
Filter AS for organic vapors
2
Filtration performances tested according to the AFNOR NF X 15-211:2009 standard : France EN 1822 : 1998 (HEPA H14 & ULPA U17 Filters) - CE Marking
100 to 120 bottles
Up to 120 x 1 L Bottles
5
Videos
SDS
Documents
Certificates
Special Offers

Special Offers

Services