missing translation for 'onlineSavingsMsg'
Learn More

Capsazepine, Tocris Bioscience™

Vanilloid receptor antagonist; also activator of ENaCδ

Brand:  Tocris (Bio-Techne) 046410

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13447713

  • £171.99 / 10mg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective vanilloid receptor antagonist (Ki = 3.2 μM). Inhibits carrageenan inflammation-induced hyperalgesic responses in the rat. Also activates amiloride-sensitive epithelial Na+ channel ENaCδ. Also available as part of the Vanilloid TRPV1 Receptor Tocriset™ .
Specifications

Chemical Identifiers

138977-28-3
376.899
capsazepine, capsazepin, n-2-4-chlorophenyl ethyl-1,3,4,5-tetrahydro-7,8-dihydroxy-2h-2-benzazepine-2-carbothioamide, unii-lfw48my844, n-2-4-chlorophenyl ethyl-7,8-dihydroxy-1,3,4,5-tetrahydro-2-benzazepine-2-carbothioamide, 2-2-4-chlorophenyl ethylamino-thiocarbonyl-7,8-dihydroxy-2,3,4,5-tetrahydro-1h-2-benzazepine, n-2-4-chlorophenyl ethyl-7,8-dihydroxy-1,3,4,5-tetrahydro-2h-2-benzazepine-2-carbothioamide, capasazepine, tocris-0464
CHEBI:70773
C1CC2=CC(=C(C=C2CN(C1)C(=S)NCCC3=CC=C(C=C3)Cl)O)O
C19H21ClN2O2S
DRCMAZOSEIMCHM-UHFFFAOYSA-N
2733484
N-[2-(4-chlorophenyl)ethyl]-7,8-dihydroxy-1,3,4,5-tetrahydro-2-benzazepine-2-carbothioamide

Specifications

10mg
138977-28-3
capsazepine, capsazepin, n-2-4-chlorophenyl ethyl-1,3,4,5-tetrahydro-7,8-dihydroxy-2h-2-benzazepine-2-carbothioamide, unii-lfw48my844, n-2-4-chlorophenyl ethyl-7,8-dihydroxy-1,3,4,5-tetrahydro-2-benzazepine-2-carbothioamide, 2-2-4-chlorophenyl ethylamino-thiocarbonyl-7,8-dihydroxy-2,3,4,5-tetrahydro-1h-2-benzazepine, n-2-4-chlorophenyl ethyl-7,8-dihydroxy-1,3,4,5-tetrahydro-2h-2-benzazepine-2-carbothioamide, capasazepine, tocris-0464
C1CC2=CC(=C(C=C2CN(C1)C(=S)NCCC3=CC=C(C=C3)Cl)O)O
376.899
CHEBI:70773
>99%
Capsazepine
C19H21ClN2O2S
DRCMAZOSEIMCHM-UHFFFAOYSA-N
N-[2-(4-chlorophenyl)ethyl]-7,8-dihydroxy-1,3,4,5-tetrahydro-2-benzazepine-2-carbothioamide
2733484
376.9
Videos
Documents