Learn More
Trajan Scientific and Medical™ BPX70 GC Capillary Column
SGE BPX70 GC Capillary Column is an MS premium, low bleed column with high polarity specifically designed for fatty acid methyl ester (FAME) analysis
Brand: Trajan Scientific and Medical™ 054623
Code : 13
Additional Details : Weight : 0.01000kg
Description
- High temperature
- Custom designed for separation of FAMEs
- Ideal for cis/trans isomer separation
- Polar phase
Specifications
GC Capillary Column | |
BPX70 | |
50°C to 250/260°C | |
60 m |
0.25 mm | |
250/260°C | |
0.25 µm |