missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CAMTA1 (Texas Red/FITC) Split FISH Probe
Labeled FISH probes for identification of gene split using Fluoresecent In Situ Hybridization Technique.
£1235.00 - £2030.00
Specifications
Content And Storage | Store at 4°C in the dark. |
---|---|
Description | Labeled FISH probes for identification of gene split using Fluoresecent In Situ Hybridization Technique. |
Format | Tube |
Label or Dye | FITC, Texas Red |
For Use With (Application) | Fluorescent In Situ Hybridization (Cell) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16013465
|
Abnova™
FS0035.200UL |
200 μL |
£2030.00
200µL |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16003465
|
Abnova™
FS0035.100UL |
100 μL |
£1235.00
100µL |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
FISH is a technique used to identify and localize the presence or absence of specific DNA sequences on cells and tissues. Abnova has developed a range of FISH probes for the detection of gene amplification, loss, and translocation. Each FISH probe has a pair of locus-specific, fluorophore-labeled probes originated from a bacterial chromosome (BAC) library.
Advantages
- Dual colored probes for fast, sensitive, and specific detection
- Work on metaphase spread, paraffin embedded, and frozen tissue
- Identify gene amplification, loss, and translocation
- High signal to noise ratio
- Low cross reactivity
Specifications
Store at 4°C in the dark. | |
Tube | |
Fluorescent In Situ Hybridization (Cell) | |
1p36.31-1p36.23/1p36.31-1p36.23 | |
CAMTA1 Split FISH Probe | |
Human |
Labeled FISH probes for identification of gene split using Fluoresecent In Situ Hybridization Technique. | |
FITC, Texas Red | |
DAPI Counterstain (1500 ng/mL ) 250 uL | |
Approx. 650kb/830kb | |
Genomic DNA | |
Liquid |