missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+/-)-Camphene, tech. (sum of camphene + fenchene), Thermo Scientific Chemicals
12.46 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
50 g
250 g
1000 g
Description
Camphene is used in the preparation of fragrances and as a food additive for flavoring.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Camphene is used in the preparation of fragrances and as a food additive for flavoring.
Solubility
Insoluble in water. Soluble in alcohol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Camphene is used in the preparation of fragrances and as a food additive for flavoring.
Solubility
Insoluble in water. Soluble in alcohol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Melting Point | ∼35°C |
Boiling Point | 159°C to 160°C |
Flash Point | 34°C (94°F) |
Quantity | 50 g |
UN Number | UN1325 |
Merck Index | 14,1730 |
Synonym | 1s-2,2-dimethyl-3-methylidenebicyclo 2.2.1 heptane, s-camphene, 1s,4r-2,2-dimethyl-3-methylene-bicyclo 2.2.1 heptane |
SMILES | CC1(C)C2CCC(C2)C1=C |
IUPAC Name | (1S,4S)-3,3-dimethyl-2-methylidenebicyclo[2.2.1]heptane |
PubChem CID | 6560154 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'