missing translation for 'onlineSavingsMsg'
Learn More

Calpeptin, Tocris Bioscience™

Calpain and cathepsin L inhibitor

Brand:  Tocris (Bio-Techne) 044850

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13437663

  • £547.32 / 50mg

Estimated Shipment
27-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent, cell-permeable inhibitor of the Ca2+-dependent protease, calpain. Prevents collagen- and thrombin-induced platelet aggregation, probably by blocking calpain induced phospholipase C and thromboxane synthase activation. Potent cathepsin L inhibitor. Recently shown to preferentially inhibit a subset of protein-tyrosine phosphatases.
Specifications

Chemical Identifiers

117591-20-5
362.47
calpeptin, n-cbz-leu-nleu-al, benzylcarbonyl-leu-nleu-h, unii-18x9fr245w, chembl92708, n-benzyloxycarbonyl-l-leucylnorleucinal, carbamic acid, 1-1-formylpentyl amino carbonyl-3-methylbutyl-, phenylmethyl ester, s-r*,r*, benzyl n-1s-3-methyl-1-2s-1-oxohexan-2-yl carbamoyl butyl carbamate, carbamic acid, 1-1-formylpentyl amino carbonyl-3-methylbutyl-, phenylemthyl ester, s-r*,r*, benzyl n-1s-3-methyl-1-2s-1-oxohexan-2-yl carbamoyl butyl carbamate
CHEBI:3330
CCCCC(C=O)NC(=O)C(CC(C)C)NC(=O)OCC1=CC=CC=C1
C20H30N2O4
PGGUOGKHUUUWAF-ROUUACIJSA-N
73364
benzyl N-[(2S)-4-methyl-1-oxo-1-[[(2S)-1-oxohexan-2-yl]amino]pentan-2-yl]carbamate

Specifications

Calpeptin
117591-20-5
calpeptin, n-cbz-leu-nleu-al, benzylcarbonyl-leu-nleu-h, unii-18x9fr245w, chembl92708, n-benzyloxycarbonyl-l-leucylnorleucinal, carbamic acid, 1-1-formylpentyl amino carbonyl-3-methylbutyl-, phenylmethyl ester, s-r*,r*, benzyl n-1s-3-methyl-1-2s-1-oxohexan-2-yl carbamoyl butyl carbamate, carbamic acid, 1-1-formylpentyl amino carbonyl-3-methylbutyl-, phenylemthyl ester, s-r*,r*, benzyl n-1s-3-methyl-1-2s-1-oxohexan-2-yl carbamoyl butyl carbamate
CCCCC(C=O)NC(=O)C(CC(C)C)NC(=O)OCC1=CC=CC=C1
362.47
CHEBI:3330
>98%
50mg
C20H30N2O4
PGGUOGKHUUUWAF-ROUUACIJSA-N
benzyl N-[(2S)-4-methyl-1-oxo-1-[[(2S)-1-oxohexan-2-yl]amino]pentan-2-yl]carbamate
73364
362.47
Videos