missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Calpeptin, ≥98%

Catalog Number p-7082234
98.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
117591-20-5
C20H30N2O4
N-Benzyloxycarbonyl-L-leucyl-norleucinal; Z-Leu-norleucinal
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
117591-20-5
C20H30N2O4
N-Benzyloxycarbonyl-L-leucyl-norleucinal; Z-Leu-norleucinal

Inhibitor of the calcium-dependent protease calpain and cathepsin L

Calpeptin is used to to inhibit the activities of cathepsin L and calpain-1. Calpeptin blocked the expression of aspartyl protease cathepsin D, amyloid precursor protein, and calparin. It acts as an inhibitor of the Ca2+-dependent protease, calpain. Prevents collagen- and thrombin-induced platelet aggregation, probably by blocking calpain induced phospholipase C and thromboxane synthase activation. Potent cathepsin L inhibitor. It is shown to preferentially inhibit a subset of protein-tyrosine phosphatases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 10 mg
Description White
Content And Storage -30°C to -10°C
For Use With (Application) Inhibitor of the calcium-dependent protease calpain and cathepsin L
Formula Weight 362.50g/mol
Solubility Soluble in DMSO at 15mg/ml
Product Type Signal Transduction Reagent
Form Powder
Assay ≥98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'