missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Calpeptin,≥98%

Inhibitor of the calcium-dependent protease calpain and cathepsin L

Brand:  Alfa Aesar™ J60481.MF

Code : NEW

Additional Details : CAS Number : 117591-20-5 Weight : 0.00050kg

 View more versions of this product

Product Code. 15453249

  • £467.80 / 50mg


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Specifications

50mg
−30° to −10°C
Inhibitor of the calcium-dependent protease calpain and cathepsin L
117591-20-5
362.50g/mol
Soluble in DMSO at 15mg/ml
White
Powder
≥98%
C20H30N2O4
N-Benzyloxycarbonyl-L-leucyl-norleucinal; Z-Leu-norleucinal
Signal Transduction Reagent
Videos
SDS