Learn More
Alfa Aesar™ Calpain Substrate I
For cellular and molecular biology applications
Brand:  Alfa Aesar™ J63222.MD
Code : NEW
Additional Details : CAS Number : 94367-20-1 Weight : 0.00025kg
 View more versions of this product
Product Code. 15495479
  • £461.31 / 250mg
Estimated Shipment date
from Supplier 12-06-2018
Add to basket
Specifications
Specifications
White
Lyophilized
Fluorogenic
Flourogenic substrate for both calpains I and II
551.59