missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calpain 15 Monoclonal Antibody (4E2), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA524564
Additional Details : Weight : 0.01000kg
Description
Peptide Sequence: HPKAYLHVQC DCTDSFNVVS TRGSLRTQDS VPPLHRQVLV ILSQLEGNAG FSITHRLAHR KAAQAFLSDW TASKGTHSPP LTPEVAGLHG PRPL
Calpain 15 Monoclonal antibody specifically detects Calpain 15 in Human samples. It is validated for ELISA, Immunocytochemistry.Specifications
Calpain 15 | |
Monoclonal | |
Unconjugated | |
CAPN15 | |
calpain 15; calpain-15; CAPN15; LOW QUALITY PROTEIN: calpain-15; small optic lobes homolog; Solh; zmp:0000001110 | |
Mouse | |
Protein A | |
RUO | |
6650 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunocytochemistry | |
4E2 | |
PBS with no preservative; pH 7.4 | |
O75808 | |
CAPN15 | |
SOLH (NP_005623, 993 a.a. ∽ 1086 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |