Learn More
Calpain 10 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA552066
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: FWLPLLEKVY AKVHGSYEHL WAGQVADALV DLTGGLAERW NLKGVAGSGG QQDRPGRWEH RTCRQLLHLK DQCLISCCVL SPRAGARELG EFHAFIVSDL Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 82%.
Calpain 10 Polyclonal antibody specifically detects Calpain 10 in Human samples. It is validated for Immunohistochemistry.Specifications
Calpain 10 | |
Polyclonal | |
Unconjugated | |
CAPN10 | |
calcium-activated neutral proteinase 10; calpain 10; calpain-10; calpain-like protease CAPN10; CANP 10; CANP10; CAPN10; KIAA1845; NIDDM1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11132 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HC96 | |
CAPN10 | |
Recombinant protein corresponding to Human Calpain 10 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only