missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ CALML5 Recombinant Human Protein, N-His.GST tag

Expressed from DNA sequence encoding human CALML5 (Met 1-Glu 146) fused to N-terminal polyhistidine-tagged GST tag at N-terminus

Brand:  Sino Biological™ 11783H20E50

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15299125

  • £370.60 / 50µg


Estimated Shipment
11-05-2018
Add to basket

Description

Description

  • Formulation: 20mM Tris, 150mM NaCl, 1mM DTT, 0.5mM GSH, 10% Glycerol, pH 7.4
Specifications

Specifications

Analysis of protein stucture and used in cell biology research applications
44.2kd
50μg
Full Length, Recombinant
Calmodulin-Like 5
Assay Standard
E.coli
GST Tag, His Tag
Lyophilized
≥90% by SDS-PAGE
Human
<1 EU/μg
CALML5
Other proteins
CALML5 Recombinant Human Protein
Videos

For Research Use Only. Not for use in diagnostic procedures.