missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calix[8]arene, 97%, Thermo Scientific Chemicals
32.26 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
Description
It is used in the study on the chromatographic behavior of water-soluble vitamins on p-tert-butyl-calix [8] arene-bonded silica gel stationary phase by HPLC.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in the study on the chromatographic behavior of water-soluble vitamins on p-tert-butyl-calix [8] arene-bonded silica gel stationary phase by HPLC.
Solubility
Sparingly soluble in water (5.8E-6 g/L) (25°C).
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It is used in the study on the chromatographic behavior of water-soluble vitamins on p-tert-butyl-calix [8] arene-bonded silica gel stationary phase by HPLC.
Solubility
Sparingly soluble in water (5.8E-6 g/L) (25°C).
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Melting Point | >450°C |
Quantity | 1 g |
Solubility Information | Sparingly soluble in water (5.8E-6g/L) (25°C). |
Formula Weight | 848.99 |
Percent Purity | 97% |
Chemical Name or Material | Calix[8]arene |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'