missing translation for 'onlineSavingsMsg'
Learn More

Calibration Buffer Solutions, Hanna Instruments™

Catalog Number p-8014376
Click to view available options
Quantity:
20 mL
pH:
4.01
10.01
This item is not returnable. View return policy
This item is not returnable. View return policy

Premium quality calibration buffer solution with reported values that are accurate to +/- 0.01 pH @ 25°C.

  • Premium quality pH 10.01 calibration buffer solution
  • 25 sachets containing 20 mL of buffer
  • Color coded label for easy pH buffer value identification
  • Accuracy of +/- 0.01 pH @ 25°C
  • Temperature chart of the actual pH value at various temperatures printed on each sachet
  • Uses high purity salts, deionized water, certified weight-checked balances, and Class A glassware
  • In a temperature controlled environment monitored with certified thermometers
  • Reported values are accurate to +/- 0.01 pH @ 25°C and are traceable to NIST Standard Reference Materials (SRMs)
  • Ideal for users that do not calibrate frequently
  • Ideal for field use due to portability
Color Colorless
pH 10.01
Physical Form Liquid
Quantity 20 mL
Solution Type Buffer
Formulation Aqueous Buffer Solution
Odor Odorless
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'