missing translation for 'onlineSavingsMsg'
Learn More

Calhex 231 hydrochloride, Tocris Bioscience™

Negative allosteric modulator of the calcium-sensing receptor

Brand:  RD Systems (Bio-Techne) 4387/10

Code : Z1

Additional Details : Weight : 0.00010kg

Quantity 10mg

 View more versions of this product

Product Code. 13464664

  • £204.60 / 10mg


Estimated Shipment
29-10-2018
Add to basket

Description

Description

Negative allosteric modulator of the calcium sensing receptor (CaSR). Blocks increases in [3H]inositol phosphate levels elicited by wild-type hCasR activation (IC50= 0.39μM in transiently transfected HEK293 cells).
Specifications

Chemical Identifiers

652973-93-8
406.954
benzamide,4-chloro-n-1s,2s-2-1r-1-1-naphthalenyl ethyl amino cyclohexyl
4-chloro-N-[(1S,2R)-2-[[(1R)-1-naphthalen-1-ylethyl]amino]cyclohexyl]benzamide
C25H27ClN2O
YTFUQWWKTIWYEY-VEYWIMSWSA-N
97103734
CC(C1=CC=CC2=CC=CC=C21)NC3CCCCC3NC(=O)C4=CC=C(C=C4)Cl

Specifications

Calhex 231 hydrochloride
652973-93-8
benzamide,4-chloro-n-1s,2s-2-1r-1-1-naphthalenyl ethyl amino cyclohexyl
CC(C1=CC=CC2=CC=CC=C21)NC3CCCCC3NC(=O)C4=CC=C(C=C4)Cl
406.954
443.41
10mg
C25H27ClN2O
YTFUQWWKTIWYEY-VEYWIMSWSA-N
4-chloro-N-[(1S,2R)-2-[[(1R)-1-naphthalen-1-ylethyl]amino]cyclohexyl]benzamide
97103734
>98%
Videos