missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium zirconium oxide, 99.2% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1 kg
Description
Calcium zirconium oxide is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 2550°C |
Physical Form | Powder |
Quantity | 250 g |
Solubility Information | Soluble in Nitric acid. |
Formula Weight | 179.3 |
Percent Purity | 99.2% |
Odor | Odorless |
Assay | (metals basis) |
Chemical Name or Material | Calcium zirconium oxide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'