missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Calcium zirconium oxide, 99.2% (metals basis)

Brand:  Alfa Aesar™ 012344.A1

Code : NEW

Additional Details : CAS Number : 12013-47-7 Weight : 1.00000kg

Quantity 1kg

 View more versions of this product

Product Code. 11301719

  • £74.00 / 1kg


Estimated Shipment date
from Supplier 24-04-2018
Add to basket

Specifications

Chemical Identifiers

Calcium zirconium oxide
179.3
Calcium zirconate
12013-47-7
MFCD00015982

Specifications

Calcium zirconium oxide
Powder
(metals basis)
MFCD00015982
−325 Mesh
2550°C
12013-47-7
99.2%
179.3
Calcium zirconate
Odorless
1kg
Videos
SDS