missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Calcium tin oxide, 99% (metals basis excluding Mg), Mg <1%

Brand:  Alfa Aesar™ 041844.22

Code : NEW

Additional Details : CAS Number : 12013-46-6 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 11344817

  • £55.60 / 100g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

Calcium tin oxide
CaSnO3
MFCD00015980
12013-46-6
206.77
Calcium stannate

Specifications

Calcium tin oxide
Powder
(metals basis excluding Mg), Mg <1%
206.77
Calcium stannate
White
12013-46-6
99%
CaSnO3
MFCD00015980
−100 Mesh
100g
Videos
SDS