Learn More
Thermo Scientific Alfa Aesar Calcium tin oxide, 99% (metals basis excluding Mg), Mg <1%
Brand: Thermo Scientific Alfa Aesar 041844.36
Calcium tin oxide, 99% (metals basis excluding Mg), Mg <1%
Code : NEW
Additional Details : CAS Number : 12013-46-6 Weight : 0.50000kg