Learn More
Calcium thiosulfate, pure, 30-50% solution in water, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 447870050
Code : 29
Additional Details : CAS Number : 10124-41-1 Weight : 6.22500kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Calcium thiosulfate | |
5 L | |
CaS2O3 | |
Solubility in water: soluble | |
[O-]S(=O)(=S)[O-].[Ca+2] | |
152.21 | |
152.21 | |
1.245 |
10124-41-1,7732-18-5 | |
CaO3S2 | |
calcium thiosulfate, tecesal, calcium thiosulphate, calcium hyposulfite, caswell no. 153, unii-2io10o728e, calcium thiosulfate ca s2o3, epa pesticide chemical code 080101, thiosulfuric acid h2s2o3 , calcium salt 1:1, thiosulfuric acid, calcium salt 1:1 | |
FAYYUXPSKDFLEC-UHFFFAOYSA-L | |
calcium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane | |
24964 | |
Pure | |
1.2450g/mL |
RUO – Research Use Only