missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium, Technical, Metal, Pieces, SLR, Fisher Chemical™
Click to view available options
Quantity:
25 g
100 g
250 g
Description
Ca, CAS Number-7440-70-2, 100g, 1484 deg.C, Gray, 40.07g/mol, 845 deg.C, 85314, Plastic powder jar, Solid
Specifications
Boiling Point | 1484°C |
Melting Point | 845°C |
Color | Gray |
pH | 14 |
Physical Form | Solid |
Quantity | 100 g |
Formula Weight | 40.07g/mol |
Packaging | Plastic powder jar |
Safety and Handling
Signal Word
- Danger
- Substance which\, in contact with water\, emits flammable gases Category 2
- H261-In contact with water releases flammable gases.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P231+P232-Handle under inert gas. Protect from moisture.
- P223-Do not allow contact with water.
- P335+P334-Brush off loose particles from skin Immerse in cool water/wrap in wet bandages.
- P370+P378-In case of fire: Use to extinguish.
- P402+P404-Store in a dry place. Store in a closed container.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'