missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium sulfate dihydrate, 98%, for biochemistry, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity | 500 g |
Assay Percent Range | 97.5% min. (Complexometry) |
Linear Formula | CaSO4·2H2O |
Solubility Information | Solubility in water: 2g/L (20°C). Other solubilities: slightly soluble in glycerol, practically insoluble in most common organic, solvents |
Formula Weight | 172.17 |
Percent Purity | 98% |
Grade | Biochemical |
Packaging | Plastic bottle |
Chemical Name or Material | Calcium sulfate dihydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'