Learn More
Invitrogen™ eBioscience™ Calcium Sensor Dye eFluor™ 514
Membrane-permeable dye used to monitor changes in intracellular free calcium concentrations in the cell. Once eFluor™ 514 Calcium Sensor Dye enters the cell, cellular esterases cleave the AM group yielding a membrane-impermeable dye fluorescing at ≈520nm.
Brand: Invitrogen™ 65-0859-39
Code : NEW
Additional Details : Weight : 0.09500kg
Description
- Calcium binding affinity: Kd = 232 nM
- Molecular Weight: ≈1100 d
- Peak Excitation: 490nm
- Peak Emission: 514nm
- Reconstitute in high-quality, freshly opened DMSO
- Recommended concentration: 2 to 5 mM
- Once reconstituted, protect from light and store dessicated at −20°C; avoid freeze-thawing
Specifications
Fluorescent | |
Fluorescence Microscope, Flow Cytometer | |
490⁄514 | |
1mg |
Calcium Assay | |
Dye | |
eBioscience™ |
For Research Use Only. Not for use in diagnostic procedures.