missing translation for 'onlineSavingsMsg'
Learn More

Calcium phosphide, Red-brown lump, Alfa Aesar™

Brand:  Alfa Aesar 019115.14

Code : NEW

Additional Details : CAS Number : 1305-99-3 Weight : 0.02500kg Transport : UN number : 1360 Chem class : 4.3 Pack group : I

Quantity 25g

 View more versions of this product

Product Code. 15481077

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

Calcium phosphide
Technical Grade
Ca3P2
MFCD00015986
1305-99-3
Lump
182.19

Specifications

Calcium phosphide
Technical Grade
Ca3P2
MFCD00015986
Moisture sensitive
25g
1305-99-3
Lump
182.19
2.51g/cm3
Red-brown
Videos
SDS