missing translation for 'onlineSavingsMsg'
Learn More

Calcium oxide, Reagent Grade, Thermo Scientific Chemicals

Catalog Number 11307618
Click to view available options
Quantity:
500 g
2 kg
1305-78-8
CaO
56.08
MFCD00010911
BRPQOXSCLDDYGP-UHFFFAOYSA-N
calcium oxide, burnt lime, calcia, calxyl, gebrannter kalk, unslaked lime, calcium monoxide, airlock, calx
14778
CHEBI:31344
oxocalcium
[O--].[Ca++]
1305-78-8
CaO
56.08
MFCD00010911
BRPQOXSCLDDYGP-UHFFFAOYSA-N
calcium oxide, burnt lime, calcia, calxyl, gebrannter kalk, unslaked lime, calcium monoxide, airlock, calx
14778
CHEBI:31344
oxocalcium
[O--].[Ca++]

Manufacturing of steel, aluminum, magnesium, and glass and calcium salts, building and construction materials, lubricants, laboratory reagentCalcium oxide has wide-ranging utility in industry. It is used in making porcelain, cements, mortars and glass. It is also employed in flue-gas desulfurization in power plants, purifying sugar, drying the solvents/reagents, preparing bleaching powder, calcium carbide and calcium cyanamide. It also finds use in water softeners, chemical recovery at Kraft pulp mills, biodiesel production and in the petroleum industry for the detection of traces of water. In agriculture, it is used to restore the pH of acidic soils (liming). It is useful as a basic catalyst and an acid scavenger in several organic transformations, for example, in Arndt-Eistert synthesis of alpha-diazoketones, synthesis of heterocycles and oximes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 2850°C
Melting Point 2572°C
Physical Form Powder
Quantity 2 kg
Solubility Information Soluble in water with generation of heat,and forming Ca(OH)2,in acids,glycerol. Insoluble in alcoholSoluble in water. Insoluble in methanol,diethyl ether and n-octanol.
Formula Weight 56.08
Grade Reagent
Refractive Index 1.83
Sensitivity Air and moisture sensitive
Density 3.25-3.38
Chemical Name or Material Calcium oxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'