missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Oxide, Powder, BAKER ANALYZED™ Reagent, J.T. Baker™
171.95 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
missing translation for 'orderingAttributeHoverText'
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Specifications
Physical Form | Powder |
Quantity | 500 g |
Formula Weight | 56.08 |
Grade | BAKER ANALYZED™ Reagent |
Packaging | Poly Bottle |
Specific Gravity | 1L = 3.37kg |
Chemical Name or Material | Calcium Oxide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'