missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Calcium oxalate, Puratronic™, 99.999% (metals basis)

Brand:  Alfa Aesar™ 046318.14

Code : NEW

Additional Details : CAS Number : 563-72-4 Weight : 0.02500kg Transport : UN number : 3288 Chem class : 1 Pack group : III

Quantity 25g

 View more versions of this product

Product Code. 15437175

  • £97.73 / 25g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

Calcium oxalate
Puratronic™
CaC2O4
MFCD00012474
563-72-4
C2CaO4
128.1
14,1685

Specifications

Calcium oxalate
Puratronic™
99.999%
C2CaO4
128.1
14,1685
Hygroscopic
25g
563-72-4
Powder
(metals basis)
CaC2O4
MFCD00012474
2.2g/mL
White
Videos
SDS