missing translation for 'onlineSavingsMsg'
Learn More

Calcium nitride, 99% (metals basis), Thermo Scientific Chemicals

Catalog Number p-4407337
Click to view available options
Quantity:
5 g
50 g
250 g
This item is not returnable. View return policy
12013-82-0
Ca3N2
MFCD00015985
This item is not returnable. View return policy
12013-82-0
Ca3N2
MFCD00015985

Calcium nitride is used in the production of light-emitting diode (LED) phosphors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 1195°C
Color Brown
Physical Form Pieces
Quantity 5 g
Assay Percent Range 99% (Metals basis)
Solubility Information Insoluble in water.
Formula Weight 148.25
Odor Ammonia-like
Sensitivity Moisture sensitive
Density 2.63
Chemical Name or Material Calcium nitride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'