missing translation for 'onlineSavingsMsg'
Learn More

Calcium Nitrate Tetrahydrate, Extra Pure, SLR, Fisher Chemical™

Catalog Number 10030050
Click to view available options
Quantity:
3 kg
This item is not returnable. View return policy
13477-34-4
CaH10N2O10
238.16
149604
DKCJOKYHYIAZQH-UHFFFAOYSA-N
calcium bis(nitric acid) tetrahydrate
O.O.O.O.[Ca++].O[N+]([O-])=O.O[N+]([O-])=O
This item is not returnable. View return policy
13477-34-4
CaH10N2O10
238.16
149604
DKCJOKYHYIAZQH-UHFFFAOYSA-N
calcium bis(nitric acid) tetrahydrate
O.O.O.O.[Ca++].O[N+]([O-])=O.O[N+]([O-])=O

Calcium Nitrate Tetrahydrate, Extra Pure, SLR

TRUSTED_SUSTAINABILITY
Boiling Point 132°C
Melting Point 45°C
Color White
pH 5 to 7
Physical Form Solid Crystalline
Quantity 3 kg
Formula Weight 236.15g/mol
Packaging Plastic powder jar
compliance-icons compliance-icons
Product Identifier
  • Calcium nitrate tetrahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'