missing translation for 'onlineSavingsMsg'
Learn More

Calcium nitrate tetrahydrate, ACS, 99.0-103.0%, Thermo Scientific Chemicals

Catalog Number 11371749
Click to view available options
Quantity:
500 g
2 kg
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
13477-34-4
CaH8N2O10
236.146
MFCD00149604
ICSSIKVYVJQJND-UHFFFAOYSA-N
calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate
16211656
CHEBI:86159
calcium;dinitrate;tetrahydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.[Ca+2]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
13477-34-4
CaH8N2O10
236.146
MFCD00149604
ICSSIKVYVJQJND-UHFFFAOYSA-N
calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate
16211656
CHEBI:86159
calcium;dinitrate;tetrahydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.[Ca+2]

Manufacturing of other nitrates, fertilizers, matches

Calcium nitrate tetrahydrate is used as nitrogenous fertilizer and corrosive inhibitor in diesel fuels. It is utilized in plant cell for culture experiments. It is also used in cooling baths and waste water handling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 132°C (decomposition)
Melting Point 42°C
Physical Form Crystalline
Quantity 2 kg
Assay Percent Range 99.0-103.0%
UN Number UN1454
Merck Index 14,1682
Solubility Information Soluble in water,alcohol,acetone and ammonia.
Formula Weight 236.15 (164.09 Anhydrous)
Percent Purity 99.0 to 103.0%
Grade ACS
Odor Odorless
Sensitivity Hygroscopic
Density 1.82 g/mL
Chemical Name or Material Calcium nitrate tetrahydrate, tetrahydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'