missing translation for 'onlineSavingsMsg'
Learn More

Calcium nitrate hydrate, Puratronic™, 99.9995% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7059399
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
35054-52-5
CaN2O6
164.09
MFCD00149604
ZCCIPPOKBCJFDN-UHFFFAOYSA-N
calcium nitrate hydrate, calcium hydrate dinitrate, acmc-20alid, calcium nitrate monohydrate, ca.2no3.h2o, nitric acid calcium salt hydrate, calcium nitrate-water 1/2/1, nitric acid, calciumsalt, hydrate 9ci
16211464
[Ca++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
This item is not returnable. View return policy
35054-52-5
CaN2O6
164.09
MFCD00149604
ZCCIPPOKBCJFDN-UHFFFAOYSA-N
calcium nitrate hydrate, calcium hydrate dinitrate, acmc-20alid, calcium nitrate monohydrate, ca.2no3.h2o, nitric acid calcium salt hydrate, calcium nitrate-water 1/2/1, nitric acid, calciumsalt, hydrate 9ci
16211464
[Ca++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 132°C (decomposition)
Melting Point 42°C
Color White
Physical Form Crystalline
Quantity 5 g
Linear Formula Ca(NO3)2·xH2O
UN Number UN1454
Merck Index 14,1682
IUPAC Name calcium dinitrate
Formula Weight 164.09 (Anhydrous)
Percent Purity 99.9995%
Grade Puratronic™
Density 1.82 g/mL
Assay (metals basis)
Chemical Name or Material Calcium nitrate hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'