missing translation for 'onlineSavingsMsg'
Learn More

Calcium Nitrate ACS AR Crystal, Macron Fine Chemicals™

Catalog Number p-3177068
180.50 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.


Click to view available options
Quantity:
500 g
Packaging:
POLYSTORMOR
This item is not returnable. View return policy
13477-34-4
CaH8N2O10
236.146
ICSSIKVYVJQJND-UHFFFAOYSA-N
calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate
16211656
CHEBI:86159
calcium;dinitrate;tetrahydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.[Ca+2]
This item is not returnable. View return policy
13477-34-4
CaH8N2O10
236.146
ICSSIKVYVJQJND-UHFFFAOYSA-N
calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate
16211656
CHEBI:86159
calcium;dinitrate;tetrahydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.[Ca+2]
Physical Form Crystalline
Quantity 500 g
Formula Weight 236.15
Grade ACS
Packaging POLYSTORMOR
Chemical Name or Material Calcium Nitrate, 4-Hydrate
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'