Calcium Ionophore A23187 (Free Acid), Fisher BioReagents

Isolated from Streptomyces chartreusensis

Overview
Brand: Fisher Bioreagents

Manufacturer Part Number: BP595-5

UNSPSC: 51101500

Code: 32

Additional Details:
Additional Details: CAS Number: 52665-69-7 Weight: 0.05001kg


Packaging Poly Micro Tube
Quantity 5mg

Product Code. 10294963

Quantity Price
1 £ 115.8 / 5mg
EU Stock 1
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Calcium Ionophore A23187
CAS 52665-69-7
Physical Form Solid
Molecular Formula C29H37N3O6
Formula Weight 523.6
Synonym Calcimycin

Specification

Formula Weight 523.6
Melting Point 182°C
Quantity 5mg
Merck Index 15, 1642
Specific Rotation -50 to -56° (+ or -)
Health Hazard 3 Emergency Overview
Harmful by inhalation, in contact with skin and if swallowed. Irritating to eyes, respiratory system and skin.
NFPA
Health:
Recommended Storage 4°C
ChemAlert Storage Symbol Gray
CAS 52665-69-7
Purity 98% min.
Carbon (C) 65.52 to 67.52%
Physical Form Solid
TLC ≥98% (purity)
Identification Pass Test
Packaging Poly Micro Tube
Molecular Formula C29H37N3O6
Health Hazard 1 Signal Word:WARNING!
Color Off-white
Synonym Calcimycin
Chemical Name or Material Calcium Ionophore A23187
Nitrogen (N) 7.82 to 8.22%
Hydrogen (H) 6.92 to 7.32%
Documentation Safety and Handling
ChemAlertStorageSymbol : Gray

Signal Word:WARNING!

Emergency Overview
Harmful by inhalation, in contact with skin and if swallowed. Irritating to eyes, respiratory system and skin.
NFPA
Health:

4°C