missing translation for 'onlineSavingsMsg'
Learn More

Calcium iodide hydrate, Thermo Scientific Chemicals

Catalog Number p-7050975
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
71626-98-7
CaI2
293.89
MFCD00149622
UNMYWSMUMWPJLR-UHFFFAOYSA-L
calcium diiodide
[Ca++].[I-].[I-]
This item is not returnable. View return policy
71626-98-7
CaI2
293.89
MFCD00149622
UNMYWSMUMWPJLR-UHFFFAOYSA-L
calcium diiodide
[Ca++].[I-].[I-]

Calcium iodide finds use in the preparation of complex used in polymerization and hydroamination as well as serving as a matrix for luminescent materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 42°C (decomposition)
Color White to Gray
Physical Form Lump
Quantity 100 g
Merck Index 14,1677
Solubility Information Freely soluble in water and alcohol
Formula Weight 293.89 (Anhydrous)
Sensitivity Hygroscopic
Density 2.55 g/mL
Chemical Name or Material Calcium iodide hydrate
compliance-icons
Product Identifier
  • Calcium iodide, hydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Calcium iodide, hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'