missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium iodide hydrate, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
Calcium iodide finds use in the preparation of complex used in polymerization and hydroamination as well as serving as a matrix for luminescent materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 42°C (decomposition) |
Color | White to Gray |
Physical Form | Lump |
Quantity | 100 g |
Merck Index | 14,1677 |
Solubility Information | Freely soluble in water and alcohol |
Formula Weight | 293.89 (Anhydrous) |
Sensitivity | Hygroscopic |
Density | 2.55 g/mL |
Chemical Name or Material | Calcium iodide hydrate |
Safety and Handling
Product Identifier
- Calcium iodide, hydrate
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : Calcium iodide, hydrate
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'