missing translation for 'onlineSavingsMsg'
Learn More

Calcium hypochlorite, tech., granular, 65% available chlorine, ACROS Organics™

Brand:  Acros Organics 300340050

Code : A0

Additional Details : CAS Number : 7778-54-3 Weight : 5.05000kg Transport : UN number : 3485 Chem class : 5.1 Pack group : II


Packaging Plastic drum
Quantity 5kg

 View more versions of this product

Product Code. 10417772

  • £92.90 / 5kg


EU Stock 7
Add to basket


Description

Description

Oxidation reagent
Specifications

Chemical Identifiers

Calcium hypochlorite
7778-54-3
≥65% (available Cl)
Ca(OCl)2
MFCD00010900
tech.
Technical Grade
CaCl2O2
142.98

Specifications

Calcium hypochlorite
7778-54-3
Technical Grade
≥65% (available Cl)
Ca(OCl)2
MFCD00010900
4.5%
1.0%
8.5%
White to off-white or faint green
5kg
tech.
others: 0.5%
Granular Solid or Granules
CaCl2O2
142.98
72%
1.0%
Plastic drum
12.50%
≈11.6 (1% aq. soln.)
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Calcium hypochlorite

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H290-May be corrosive to metals.
  • H272-May intensify fire; oxidiser.
  • EUH031-Contact with acids liberates toxic gas.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS