missing translation for 'onlineSavingsMsg'
Learn More

Calcium hydroxide, ACS, 95.0% min, Thermo Scientific Chemicals

Catalog Number p-7049550
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy
1305-62-0
CaH2O2
74.09
MFCD00010901
AXCZMVOFGPJBDE-UHFFFAOYSA-L
calcium hydroxide, slaked lime, hydrated lime, calcium dihydroxide, calcium ii hydroxide, calcium oxide, hydrated, kalziumhydroxid, loeschkalk, pickling lime, calciumdihydroxid
6093208
CHEBI:31341
[OH-].[OH-].[Ca++]
This item is not returnable. View return policy
1305-62-0
CaH2O2
74.09
MFCD00010901
AXCZMVOFGPJBDE-UHFFFAOYSA-L
calcium hydroxide, slaked lime, hydrated lime, calcium dihydroxide, calcium ii hydroxide, calcium oxide, hydrated, kalziumhydroxid, loeschkalk, pickling lime, calciumdihydroxid
6093208
CHEBI:31341
[OH-].[OH-].[Ca++]

Air Sensitive

Mortar, plasters, cements, gas manufacturing to remove ammonia, in lubricants, water softening, petrochemicals, soil conditioner, paving materialsCalcium hydroxide is used as a flocculant in water and sewage treatment. It is also used in fresh water treatment to raise the pH of the water thereby controlling the corrosion of pipes. It is widely used in the production of calcium stearate. It is an active ingredient in whitewash, mortar and plaster. This compound is also used in the petroleum refining industry for the preparation of additives to oils and in the preparation of dry mixes for painting and decorating. It finds application as an antifungal and antimicrobial preservative for vegetables in storage.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Mortar, plasters, cements, gas manufacturing to remove ammonia, in lubricants, water softening, petrochemicals, soil conditioner, paving materials
TRUSTED_SUSTAINABILITY
Physical Form Powder
Quantity 250 g
Assay Percent Range 95.0% min.
Linear Formula Ca(OH)2
UN Number UN3262
Merck Index 14,1673
Solubility Information Slightly soluble in water; Soluble in glycerol,acids; Insoluble in alcohol.
IUPAC Name calcium;dihydroxide
Formula Weight 74.09
Percent Purity ≥95.0%
Grade ACS
Odor Odorless
Sensitivity Air Sensitive
Appearance White powder
Density 2.24 g/cm3
Chemical Name or Material Calcium hydroxide, For ACS analysis
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'