missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium hydrogen phosphate, anhydrous, 98% min, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1 kg
5 kg
Description
It is used as a feed additive for livestock and poultry. It can promote feed digestion, put on animal’s weight, raise laying rate or producing milk rate, cure diseases such as rickets, osteomalacia and anemia.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White |
Physical Form | Powder |
Quantity | 1 kg |
Linear Formula | CaHPO4 |
Merck Index | 14,1692 |
Solubility Information | Soluble in hydrochloric acid,nitric acid,acetic acid,ammonium citrate solution. Slightly soluble in water. Insoluble in ethanol. |
Formula Weight | 136.06 |
Percent Purity | ≤98% |
Odor | Odorless |
Chemical Name or Material | Calcium hydrogen phosphate, Anhydrous |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'