missing translation for 'onlineSavingsMsg'
Learn More

Calcium granules, redistilled, -6 mesh, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog Number 11306677
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7440-70-2
Ca
40.08
MFCD00010897 MFCD00085314
OYPRJOBELJOOCE-UHFFFAOYSA-N
unii-sy7q814vup, hsdb 273, sy7q814vup, compounds, elemental, calcium, elemental, kalzium, atom, ingot, chelate
5460341
CHEBI:29320
calcium
[Ca]
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7440-70-2
Ca
40.08
MFCD00010897 MFCD00085314
OYPRJOBELJOOCE-UHFFFAOYSA-N
unii-sy7q814vup, hsdb 273, sy7q814vup, compounds, elemental, calcium, elemental, kalzium, atom, ingot, chelate
5460341
CHEBI:29320
calcium
[Ca]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Silver-White
Appearance Silvery-white granules
Density 1.54g/cm3
Assay Percent Range 99.5% (Metals basis)
Quantity 250 g
Mesh Size -6 Mesh
Odor Odorless
Form Granular
Chemical Name or Material Calcium, Redistilled
compliance-icons
Product Identifier
  • Calcium granules
Signal Word
  • Danger
Hazard Category
  • Substance which\, in contact with water\, emits flammable gases Category 2
Hazard Statement
  • H261-In contact with water releases flammable gases.
Precautionary Statement
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P233-Keep container tightly closed.
  • P370+P378-In case of fire: Use to extinguish.
  • P402+P404-Store in a dry place. Store in a closed container.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'