missing translation for 'onlineSavingsMsg'
Learn More

Calcium fluoride, Reagent Grade, 97%, Thermo Scientific Chemicals

Catalog Number p-4781837
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
7789-75-5
CaF2
78.08
MFCD00010907
WUKWITHWXAAZEY-UHFFFAOYSA-L
calcium fluoride, kalziumfluorid, kalziumdifluorid, calcium 2+ difluoride, acmc-1bk0c, ksc377c5h, calcium fluoride 100g, calcium fluoride,optical grade
84512
CHEBI:35437
[F-].[F-].[Ca++]
This item is not returnable. View return policy
7789-75-5
CaF2
78.08
MFCD00010907
WUKWITHWXAAZEY-UHFFFAOYSA-L
calcium fluoride, kalziumfluorid, kalziumdifluorid, calcium 2+ difluoride, acmc-1bk0c, ksc377c5h, calcium fluoride 100g, calcium fluoride,optical grade
84512
CHEBI:35437
[F-].[F-].[Ca++]

Calcium fluoride is used to manufacture optical components such as windows and lenses, used in thermal imaging systems, spectroscopy, and excimer lasers, electronics, lasers, high-temperature dry-film lubricants, emery wheels, certain cements, phosphors, paint pigment, catalyst in wood preservatives, and optical equipment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point ca 2500°C
Melting Point 1403°C
Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range 97%
Solubility Information Insoluble in water. Soluble in ammonium salts. Soluble in conc. mineral acids with evolution of HF
IUPAC Name calcium;difluoride
Formula Weight 78.08
Grade Reagent
Odor Odorless
Refractive Index 1.434
Sensitivity Hygroscopic
Density 3.18 g/cm3 at 20°C
Chemical Name or Material Calcium fluoride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'