missing translation for 'onlineSavingsMsg'
Learn More

Calcium fluoride, Vacuum Deposition Grade, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7050279
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
7789-75-5
CaF2
78.08
MFCD00010907
WUKWITHWXAAZEY-UHFFFAOYSA-L
calcium fluoride, kalziumfluorid, kalziumdifluorid, calcium 2+ difluoride, acmc-1bk0c, ksc377c5h, calcium fluoride 100g, calcium fluoride,optical grade
84512
CHEBI:35437
[F-].[F-].[Ca++]
This item is not returnable. View return policy
7789-75-5
CaF2
78.08
MFCD00010907
WUKWITHWXAAZEY-UHFFFAOYSA-L
calcium fluoride, kalziumfluorid, kalziumdifluorid, calcium 2+ difluoride, acmc-1bk0c, ksc377c5h, calcium fluoride 100g, calcium fluoride,optical grade
84512
CHEBI:35437
[F-].[F-].[Ca++]

Calcium fluoride is used to produce a wide range of materials, manufacture optical components such as windows and lenses, used in thermal imaging systems, spectroscopy, and excimer lasers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point ∼2500°C
Melting Point 1403°C
Physical Form Melted Pieces
Quantity 10 g
Merck Index 14,1667
Solubility Information Slightly soluble in acid. Insoluble in acetone.
IUPAC Name calcium;difluoride
Formula Weight 78.08
Percent Purity 99.9%
Odor Odorless
Refractive Index 1.434
Density 3.18 g/mL
Assay (metals basis)
Chemical Name or Material Calcium fluoride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'