missing translation for 'onlineSavingsMsg'
Learn More

Calcium fluoride crystal optic rectangle, 38.5mm x 19.5mm x 4mm (drilled), polished both sides, Thermo Scientific Chemicals

 

Brand:  Thermo Scientific Alfa Aesar 038476.KT

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15493655

  • £178.00 / Each
Estimated Shipment: 24-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Calcium fluoride crystal optic rectangle
38.5mm x 19.5mm x 4mm (drilled), Polished both sides
1 Ea.
MFCD00010907
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only