missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Calcium fluoride crystal optic disc, 13mm x 2mm, polished both sides

Brand:  Alfa Aesar™ 038428.KT

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15466225

  • £77.40 / Each


Estimated Shipment date
from Supplier 22-01-2018
Add to basket

Specifications

Specifications

Calcium fluoride crystal optic disc
13mm x 2mm, Polished both sides
MFCD00010907
1 each
Videos
SDS