missing translation for 'onlineSavingsMsg'
Learn More

Calcium fluoride, 99.95% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7048603
50.13 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7789-75-5
CaF2
78.08
MFCD00010907
WUKWITHWXAAZEY-UHFFFAOYSA-L
calcium fluoride, kalziumfluorid, kalziumdifluorid, calcium 2+ difluoride, acmc-1bk0c, ksc377c5h, calcium fluoride 100g, calcium fluoride,optical grade
84512
CHEBI:35437
[F-].[F-].[Ca++]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7789-75-5
CaF2
78.08
MFCD00010907
WUKWITHWXAAZEY-UHFFFAOYSA-L
calcium fluoride, kalziumfluorid, kalziumdifluorid, calcium 2+ difluoride, acmc-1bk0c, ksc377c5h, calcium fluoride 100g, calcium fluoride,optical grade
84512
CHEBI:35437
[F-].[F-].[Ca++]

Calcium fluoride is the commercially important source for fluoride compounds as well as hydrogen fluoride. It is transparent from ultraviolet to infrared region. It is used in spectroscopy, thermal imaging systems and excimer lasers; it is also used in the manufacturing of optical components including windows, prisms and lenses. In addition, it is also used in aluminum-metallurgy, brake lining manufacturing industries, and in tooth enamel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point ∼2500°C
Melting Point 1403°C
Physical Form Powder
Quantity 25 g
Beilstein EM>5,
Merck Index 14,1667
Solubility Information Slightly soluble in acid; insoluble in acetone.
IUPAC Name calcium difluoride
Formula Weight 78.08
Percent Purity 99.95%
Odor Odorless
Refractive Index 1.434
Sensitivity Hygroscopic
Density 3.18 g/mL
Assay (metals basis)
Chemical Name or Material Calcium fluoride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'