missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium fluoride, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
It finds its application in spectroscopy, electronics, lasers, high-temperature dry-film lubricants, emery wheels, certain cements, phosphors, paint pigment, catalyst in wood preservatives, and optical equipment. Calcium fluoride is used as component of electrolyte, fluxing agent for aluminum metallurgy. It is used in glass and fluorescent lamps industry. It has dental applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | ∼2500°C |
Melting Point | 1403°C |
Physical Form | Crystals |
Quantity | 250 g |
Merck Index | 14,1667 |
Solubility Information | Soluble in ammonium salts. Insoluble in water. |
Formula Weight | 78.08 |
Percent Purity | 99% |
Odor | Odorless |
Refractive Index | 1.434 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'