missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium D-gluconate monohydrate, 98+%, Thermo Scientific Chemicals
11.60 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
2500 g
Description
Calcium D-gluconate monohydrate is used as a soluble for developing the highly effective, durable, and cheap CaO-based sorbents suitable for CO2 capture, precursor for CaO-based sorbents, in medicine, treatment of hypocalcemia, as an antidote.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Calcium D-gluconate monohydrate is used as a soluble for developing the highly effective, durable, and cheap CaO-based sorbents suitable for CO2 capture, precursor for CaO-based sorbents, in medicine, treatment of hypocalcemia, as an antidote.
Solubility
Soluble in water(30g/L).
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Calcium D-gluconate monohydrate is used as a soluble for developing the highly effective, durable, and cheap CaO-based sorbents suitable for CO2 capture, precursor for CaO-based sorbents, in medicine, treatment of hypocalcemia, as an antidote.
Solubility
Soluble in water(30g/L).
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications
Melting Point | ∽195°C (decomposition) |
Assay Percent Range | ≥98% |
Molecular Formula | C12H22CaO14 |
Quantity | 100 g |
Merck Index | 14,1670 |
Synonym | calcium gluconate, calcium d-gluconate, calciofon, calglucon, glucobiogen, ebucin, calcicol, calcipur, calglucol, dragocal |
Solubility Information | Soluble in water(30g/L). |
InChI Key | NEEHYRZPVYRGPP-IYEMJOQQSA-L |
SMILES | C(C(C(C(C(C(=O)[O-])O)O)O)O)O.C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[Ca+2] |
IUPAC Name | calcium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'