missing translation for 'onlineSavingsMsg'
Learn More

Calcium cyanamide, tech., Thermo Scientific Chemicals

Catalog Number p-7080510
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
156-62-7
CCaN2
MFCD00064894
This item is not returnable. View return policy
156-62-7
CCaN2
MFCD00064894

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 1340°C
Color Gray
Physical Form Powder
Quantity 100 g
Linear Formula CaNCN
UN Number UN1403
Beilstein 4124391
Merck Index 14,1662
Formula Weight 80.11
Grade Technical
Odor Specific type
Sensitivity Moisture sensitive
Density 2.29 g/mL
Chemical Name or Material Calcium cyanamide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'