missing translation for 'onlineSavingsMsg'
Learn More

Calcium citrate tribasic tetrahydrate, 98+%, for analysis, Thermo Scientific Chemicals

Catalog Number p-3593393
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
5785-44-4
C12H10Ca3O14·4H2O
570.5
MFCD00150786
LNIZKKFWMDARJV-UHFFFAOYSA-H
calcium citrate tetrahydrate, unii-mlm29u2x85, calcium citrate usp, tricalcium dicitrate tetrahydrate, citric acid calcium salt, citracal tn, calcium citrate 3:2 , tetrahydrate, acmc-20aliq, ksc493e4f
5282392
tricalcium;2-hydroxypropane-1,2,3-tricarboxylate;tetrahydrate
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.O.O.[Ca+2].[Ca+2].[Ca+2]
This item is not returnable. View return policy
5785-44-4
C12H10Ca3O14·4H2O
570.5
MFCD00150786
LNIZKKFWMDARJV-UHFFFAOYSA-H
calcium citrate tetrahydrate, unii-mlm29u2x85, calcium citrate usp, tricalcium dicitrate tetrahydrate, citric acid calcium salt, citracal tn, calcium citrate 3:2 , tetrahydrate, acmc-20aliq, ksc493e4f
5282392
tricalcium;2-hydroxypropane-1,2,3-tricarboxylate;tetrahydrate
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.O.O.[Ca+2].[Ca+2].[Ca+2]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Loss on Drying 10.0 % to 13.3 % (150°C, 4 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98.0 to 102% (on dried basis) (Complexometry)
Packaging Plastic bottle
Quantity 250 g
Merck Index 15, 1663
Solubility Information Solubility in water: 1g/L (20°C)
Formula Weight 570.5
Percent Purity 98+%
Grade Analysis
Insoluble Matter 0.2% max. (in HCl)
Physical Form Liquid
Chemical Name or Material Calcium citrate tribasic tetrahydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'